missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBLAC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566316
This item is not returnable.
View return policy
Description
Immunogen sequence: MSALEWYAHKS LGDGIFWIQE RFYESGNRAN IWLVRGSEQD VVIDTGLGLR SLPEYLYSSG LLQDREAKED A Highest antigen sequence identity to the following orthologs - mouse 96%, rat 97%.
MBLAC2 is a protein coding gene.
Specifications
| MBLAC2 | |
| Polyclonal | |
| Unconjugated | |
| MBLAC2 | |
| MBLAC2; metallo-beta-lactamase domain containing 2; metallo-beta-lactamase domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 153364 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68D91 | |
| MBLAC2 | |
| Recombinant protein fragment of Human MBLAC2 (Product #RP-106995) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction