missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBLAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571151
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Rat: 86%.
MBLAC1 gene ontology annotations related to this gene include hydrolase activity; metal ion binding.
Specifications
| MBLAC1 | |
| Polyclonal | |
| Unconjugated | |
| MBLAC1 | |
| MBLAC1; metallo-beta-lactamase domain containing 1; metallo-beta-lactamase domain-containing protein 1; RGD1306932; Unknown (protein for MGC:137924) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 255374 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A4D2B0 | |
| MBLAC1 | |
| synthetic peptide directed towards the C-terminal region of human MBLAC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction