missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBL2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £361.00
Specifications
| Antigen | MBL2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17918862
|
Invitrogen™
PA5145328 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17969522
![]() |
Invitrogen™
PA5145327 |
200 μL |
£361.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.Specifications
| MBL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chicken, Rat | |
| P08661, Q98TA4 | |
| 374267, 64668 | |
| Recombinant Chicken MBL protein, Leu22-Leu254 (Accession #Q98TA4). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 27 kDa mannan-binding protein monomeric subunit; Ab2-001; Ab2-011; cMBl; COLEC1; collectin; Collectin1; collectin-1; c-type lectin; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding lectin C; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose binding lectin, liver (C); mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin precursor protein; mannose-binding lectin-C; mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); mbl; MBL2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; pMBP-27; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin | |
| MBL2 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| MBL2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title