Läs mer
Invitrogen™ MBL2 Monoclonal Antibody (3E7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA140145
Beskrivning
MA1-40145 detects MBL from human samples. The MA1-40145 immunogen is Mannose Binding Lectin (MBL). MA1-40145 has been successfully used in flow cytometry, Western blot, ELISA, Functional Assays, and immunohistochemistry (frozen) applications. Negative control: Nonimmune normal serum or TBS. Endotoxin <24 EU/mg.
MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
Specifikationer
| MBL2 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% BSA and no preservative | |
| P11226 | |
| MBL2 | |
| Serum isolated human RaRF. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
| 3E7 | |
| Unconjugated | |
| MBL2 | |
| 27 kDa mannan-binding protein monomeric subunit; Ab2-001; Ab2-011; cMBl; COLEC1; collectin; Collectin1; collectin-1; c-type lectin; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding lectin C; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose binding lectin, liver (C); mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin precursor protein; mannose-binding lectin-C; mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); mbl; MBL2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; pMBP-27; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin | |
| Mouse | |
| Protein G | |
| RUO | |
| 4153 | |
| 4°C | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.