Learn More
MBL™ Borosilicate Glass Class A Col Lisse
Calibrated to Class A tolerances. MBL™ Borosilicate Glass Class A Col Lisse is iIndividually laser etched with serial number, batch code and date of manufacture
Brand:  MBL FRD206
Code : A0
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 12200353
  • £48.00 / Pack of 5
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
  • Complies with ISO 1042 and DIN 12664
  • Finely calibrated to Class A tolerances
  • Each flask is laser etched with an individual serial number, batch code and date of manufacture
  • Manufactured from chemically resistant borosilicate glass
  • Conformity 'H' certified
  • Inscriptions in blue enamel
  • Supplied with polyethylene stopper
  • Batch certificate available to download online
  • Complies with ISO 1042
  • With plain neck to accept rubber stopper
  • Manufactured from borosilicate glass
  • Finely calibrated to Class A tolerances
  • Conformity 'H' certified
  • Individually laser etched with serial number, batch code and date of manufacture
  • Batch certificate available to download online
  • Blue enamel inscriptions
Specifications
Specifications
Class A, ISO 1042, Conformity 'H' certified
25 mL
110 mm
±0.04 mL
A
Rubber
Borosilicate Glass
Plain
Yes