missing translation for 'onlineSavingsMsg'
Learn More

MBD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18347354 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347354 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347354 Supplier Novus Biologicals Supplier No. NBP310065100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MBD4 Polyclonal specifically detects MBD4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human MBD4 (NP_001263199.1). Peptide sequence NNCSPTRKDFTGEKIFQEDTIPRTQIERRKTSLYFSSKYNKEALSPPRRK
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8930
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.