missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | MBD4 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MBD4 Polyclonal specifically detects MBD4 in Human samples. It is validated for Western Blot.Specifications
| MBD4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics | |
| PBS buffer, 2% sucrose | |
| 8930 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein | |
| The immunogen is a synthetic peptide directed towards the middle region of human MBD4 (NP_001263199.1). Peptide sequence TSLKPEDFDFTVLSKRGIKSRYKDCSMAALTSHLQNQSNNSNWNLRTRSK | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title