missing translation for 'onlineSavingsMsg'
Learn More

MBD4 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18365280 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365280 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365280 Supplier Novus Biologicals Supplier No. NBP191189JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MBD4 Polyclonal specifically detects MBD4 in Human, Mouse (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD4
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein
Gene Symbols MBD4
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human MBD4 protein (between residues 200-250) [UniProt O95243]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8930.0
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.