missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBD3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148763
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. Necessary for centrosome duplication. It functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation.
Specifications
| MBD3 | |
| Polyclonal | |
| Unconjugated | |
| MBD3 | |
| AI181826; AU019209; CpG binding domain 3; Mbd3; methyl-CpG binding domain protein 3; methyl-CpG-binding domain protein 3; Methyl-CpG-binding protein MBD3 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 53615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O95983 | |
| MBD3 | |
| E. coli-derived recombinant human MBD3 isoform 2, Met1-Val259. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction