missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MBD3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17742204
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17742204 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17742204 Supplier Invitrogen™ Supplier No. PA5120806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, K-562. The target is usually found in the following locations: Chromosome, Nucleus. Immunogen sequence: MERKSPSGKK FRSKPQLARY LGGSMDLSTF DFRTGKMLMS KMNKSRQRVR YDSSNQVKGK PDLNTALPVR QTASIFKQPV TKITNHPSNK VKSDPQKAVD QPRQLFWEKK LSGLNAFDIA EELVKTMDLP KGLQGVGPGC TDETLLSAIA SALHTSTMPI TGQLSAAVEK NPGVWLNTTQ PLCKAFMVTD EDIRKQEELV QQVRKRLEEA LMADMLAHVE ELARDGEAPL DKACAEDDDE EDEEEEEEEP DPDPE.

Regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. Necessary for centrosome duplication. It functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MBD3
Gene Accession No. O95983, Q9Z2D8
Gene Alias AI181826; AU019209; CpG binding domain 3; Mbd3; methyl-CpG binding domain protein 3; methyl-CpG-binding domain protein 3; Methyl-CpG-binding protein MBD3
Gene Symbols MBD3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human MBD3 (NP_001268383.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17192, 53615
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.