missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD3 Antibody (PCRP-MBD3-1C4), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08535AF405
This item is not returnable.
View return policy
Description
MBD3 Monoclonal antibody specifically detects MBD3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| MBD3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| methyl-CpG binding domain protein 3, methyl-CpG-binding domain protein 3, Methyl-CpG-binding protein MBD3 | |
| Recombinant full-length human MBD3 protein (Uniprot: O95983) | |
| 100 μg | |
| Chromatin Research | |
| 53615 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-MBD3-1C4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction