missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBD2 Recombinant Rabbit Monoclonal Antibody (24GB460)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | MBD2 |
|---|---|
| Clone | 24GB460 |
| Concentration | 0.8 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275516
![]() |
Invitrogen™
MA557472 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276250
|
Invitrogen™
MA557473 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBD2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Heterochromatin protein-1 (HP1) is a methyl-lysine binding protein localized at heterochromatin sites, where it mediates gene silencing. It has been shown that mammalian methyltransferases that selectively methylate histone H3 on lysine-9 generate a binding site for HP1 proteins, a family of heterochromatic adaptor molecules implicated in both gene silencing and supranucleosomal chromatin structure. High-affinity in vitro recognition of a methylated histone H3 peptide by HP1 requires a functional chromodomain. Thus, the HP1 chromodomain is a specific interaction motif for the methyl epitope on lysine-9 of histone H3. In vivo, heterochromatin association of HP1 proteins is lost in Suv39h double-null primary mouse fibroblasts but is restored after reintroduction of a catalytically active SUV39H1 HMTase.Specifications
| MBD2 | |
| 0.8 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBB5, Q9Z2E1 | |
| 17191, 680172, 8932 | |
| A synthesized peptide derived from human MBD2 (aa 100-165). | |
| Antibody |
| 24GB460 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Mbd2 | |
| demethylase; DKFZp586O0821; DMTase; MBD2; MBD2a; methyl-CpG binding domain protein 2; methyl-CpG-binding domain protein 2; Methyl-CpG-binding protein MBD2; NY-CO-41 | |
| Mbd2 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title