missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99402DL594
This item is not returnable.
View return policy
Description
MBD2 Polyclonal antibody specifically detects MBD2 in Human samples. It is validated for Western Blot
Specifications
| MBD2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| demethylase, DKFZp586O0821, DMTase, methyl-CpG binding domain protein 2, methyl-CpG-binding domain protein 2, Methyl-CpG-binding protein MBD2, NY-CO-41 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MBD2. | |
| 0.1 mL | |
| Breast Cancer, Chromatin Research, Stem Cell Markers | |
| 8932 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction