missing translation for 'onlineSavingsMsg'
Learn More

MBD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18360543
Change view
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
25µL
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360543 50 μg 50µg
18333685 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18360543 Supplier Bio-Techne Supplier No. NBP31867450UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MBD1 Polyclonal antibody specifically detects MBD1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.6 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000
Formulation PBS
Gene Accession No. Q9UIS9
Gene Alias CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human MBD1 (Methyl-CpG-binding domain protein 1), using 3 KLH-conjugated synthetic peptides, two containing a sequence from the C-terminal part and one containing a sequence from the N-terminal part of the protein.
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.