missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD1 Polyclonal Antibody
MBD1 Polyclonal Antibody
Brand: Invitrogen PA1889
This item is not returnable.
View return policy
Description
MBD1 Polyclonal Antibody for Western Blot
Specifications
| MBD1 | |
| Polyclonal | |
| Unconjugated | |
| MBD1 | |
| CXXC3; CXXC-type zinc finger protein 3; Mbd1; methyl-CpG binding domain protein 1; methyl-CpG binding domain protein 1 isoform PCM1; methyl-CpG-binding domain protein 1; Methyl-CpG-binding protein MBD1; PCM1; Protein containing methyl-CpG-binding domain 1; RFT; the regulator of fibroblast growth factor 2 (FGF-2) transcription | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4152 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Q9UIS9 | |
| MBD1 | |
| Synthetic Peptide: C R(593) S K D L K K P G A R K Q(605). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction