missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD1 Antibody (PSH01-78), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32572
This item is not returnable.
View return policy
Description
MBD1 Monoclonal antibody specifically detects MBD1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| MBD1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription | |
| Recombinant protein within mouse MBD1 aa 1-200/ 636. (Uniprot: Q9UIS9) | |
| 100 μL | |
| Chromatin Research | |
| 4152 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| PSH01-78 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction