missing translation for 'onlineSavingsMsg'
Learn More

MBD1 Antibody (100B272.1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 8 publications

£189.00 - £418.00

Specifications

Antigen MBD1
Clone 100B272.1
Concentration 0.5 mg/mL
Dilution Western Blot 2-4 ug/ml, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 25798578), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24464130), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 15327775), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24464130), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18021944
View Documents
Novus Biologicals
NB100-56537SS
0.025 mg
£189.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18745143
View Documents
Novus Biologicals
NB100-56537
0.1 mg
£418.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MBD1 Monoclonal specifically detects MBD1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Specifications

Specifications

MBD1
0.5 mg/mL
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown
Unconjugated
Mouse
Chromatin Research
Q9UIS9
4152
This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 391-405 (SESEDGAGSPPPYRR) of human MBD1; GenBank no ref NP_056671.2
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
100B272.1
Western Blot 2-4 ug/ml, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 25798578), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24464130), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 15327775), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24464130), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated
Monoclonal
Purified
RUO
Human, Mouse
CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
MBD1
IgG1
Protein G purified
There are many isoforms of human MBD1 ranging from 503 (∼55 kDa) to 655 (∼71 kDa) amino acids in length. Researchers are encouraged to use the provided immunogen sequence to determine if this antibody is suitable for testing a specific isoform of interest.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.