missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MB 05032

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MB 05032 is a potent FBPase inhibitor (EC50 values are 1.6, 29 and 61 nM for human liver FBPase, human muscle FBPase and rat liver FBPase). Promotes hematopoietic stem cell and progenitor cell expansion. Increases engraftment in primary NSG mice recipients. Also enhances myeloid and lymphoid chimerism in secondary NSG mouse recipients. Promotes glycolysis, glucose uptake and lactate production in umbilical cord blood CD34+ cells.
Specifications
Specifications
| Chemical Name or Material | P-[5-[2-Amino-5-(2-methylpropyl)-4-thiazolyl]-2-furanyl]phosphonic acid |
| CAS | 261365-11-1 |
| Quantity | 10 mg |
| Molecular Formula | C11H15N2O4PS |
| Formula Weight | 302.29 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction