missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MB 05032
GREENER_CHOICE

Product Code. 18776796
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18776796

Brand: Tocris Bioscience™ 6618/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent FBPase inhibitor; promotes HSC expansion

MB 05032 is a potent FBPase inhibitor (EC50 values are 1.6, 29 and 61 nM for human liver FBPase, human muscle FBPase and rat liver FBPase). Promotes hematopoietic stem cell and progenitor cell expansion. Increases engraftment in primary NSG mice recipients. Also enhances myeloid and lymphoid chimerism in secondary NSG mouse recipients. Promotes glycolysis, glucose uptake and lactate production in umbilical cord blood CD34+ cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material P-[5-[2-Amino-5-(2-methylpropyl)-4-thiazolyl]-2-furanyl]phosphonic acid
CAS 261365-11-1
Quantity 50 mg
Molecular Formula C11H15N2O4PS
Formula Weight 302.29
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.