missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAZ Polyclonal Antibody
GREENER_CHOICE

Product Code. 15524802
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15524802 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15524802 Supplier Invitrogen™ Supplier No. PA541434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: FPVFPCTLLA PPFPVLGLDS RGVGGLMNSF PPPQGHAQNP LQVGAELQSR Sequence homology: Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors. Regulates inflammation-induced expression of serum amyloid A proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAZ
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAZ
Gene Accession No. P56270, P56671
Gene Alias LOW QUALITY PROTEIN: myc-associated zinc finger protein; Maz; MAZI; MYC associated zinc finger protein; myc-associated zinc finger protein; MYC-associated zinc finger protein (purine-binding transcription factor); PUR1; Pur-1; purine-binding transcription factor; SAF-1; SAF-2; SAF-3; serum amyloid A activating factor 1; serum amyloid A activating factor 2; serum amyloid A-activating factor-1; Transcription factor Zif87; ZF87; Zif87; Zinc finger protein 801; zinc-finger protein, 87 kilodaltons; ZNF801
Gene Symbols MAZ
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MAZ (aa 2-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17188, 4150
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.