missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MAZ Antibody [PE], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NB100-86984PE

Product Code. 30028487

  • £467.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MAZ Polyclonal antibody specifically detects MAZ in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),ChIP assay (ChIP),KnockDown
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAZ
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
4150
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), ChIP Assay, KnockDown
PE
MAZI, myc-associated zinc finger protein, MYC-associated zinc finger protein (purine-binding transcription factor), PUR1, Pur-1SAF-2, Purine-binding transcription factor, serum amyloid A activating factor 1, serum amyloid A activating factor 2, Transcription factor Zif87, ZF87SAF-1, Zif87, Zinc finger protein 801, zinc-finger protein, 87 kilodaltons, ZNF801SAF-3
The immunogen recognized by this antibody maps to a region between residue 427 and 477 of human MYC-associated zinc finger protein using the numbering given in entry BAA33064.1 (GeneID 4150).
0.1 mL
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.