missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAZ Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-86984B
This item is not returnable.
View return policy
Description
MAZ Polyclonal antibody specifically detects MAZ in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MAZ | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| MAZI, myc-associated zinc finger protein, MYC-associated zinc finger protein (purine-binding transcription factor), PUR1, Pur-1SAF-2, Purine-binding transcription factor, serum amyloid A activating factor 1, serum amyloid A activating factor 2, Transcription factor Zif87, ZF87SAF-1, Zif87, Zinc finger protein 801, zinc-finger protein, 87 kilodaltons, ZNF801SAF-3 | |
| The immunogen recognized by this antibody maps to a region between residue 427 and 477 of human MYC-associated zinc finger protein using the numbering given in entry BAA33064.1 (GeneID 4150). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4150 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction