missing translation for 'onlineSavingsMsg'
Learn More

Maytansine, TRC

Product Code. 30133072
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133072 10 mg 10mg
30004999 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133072 Supplier Toronto Research Chemicals Supplier No. TRCM19794010MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35846-53-8
Molecular Formula C34H46ClN3O10
Molecular Weight (g/mol) 692.2
Synonym MTS, Maitansine, Maysanine, Maytansin, N-Acetyl-N-methyl-L-alanine [1S-(1R*,2S*,3R*,5R*,6R*,16E,18E,20S*,21R*)]-11-Chloro-21-hydroxy-12,20-dimethoxy-2,5,9,16-tetramethyl-8,23-dioxo-4,24-dioxa-9,22-diazatetracyclo[19.3.1.110,14.03,5]hexacosa-10,12,14(26),16,18-pentaen-6-yl Ester, NSC 153858
SMILES [NH4+].[NH4+].[NH4+].NC1=NC(=O)N(C=C1)[C@H]2C(F)(F)C(O)[C@@H](COP([O-])(=O)OP([O-])(=O)[O-])O2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C34H46ClN3O10/c1-18-11-10-12-26(45-9)34(43)17-25(46-32(42)36-34)19(2)30-33(5,48-30)27(47-31(41)20(3)37(6)21(4)39)16-28(40)38(7)23-14-22(13-18)15-24(44-8)29(23)35/h10-12,14-15,19-20,25-27,30,43H,13,16-17H2,1-9H3,(H,36,42)/b12-10+,18-11+/t19-,20+,25+,26-,27+,30+,33+,34+/m1/s1
Percent Purity >95%
Chemical Name or Material Maytansine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.