missing translation for 'onlineSavingsMsg'
Learn More

Maybridge™ Maybridge HitCreator Screening Library

Product Code. 18846156
Change view
Click to view available options
Content And Storage:
45 Plates, 0.25 μmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage
18846156 45 Plates, 0.25 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18846156 Supplier Maybridge™ Supplier No. HC020384
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

13,800 compounds conveniently supplied as dry films in Thermo Scientific Matrix 96 shallow-well plates

HitCreator is derived from a rigorous analysis of more than 550,000 commercially available screening compounds to offer exceptional diversity for such a small library.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Screening Compound Library
Plate Material Matrix plate
For Use With (Application) High Throughput Screening
Content And Storage 45 Plates, 0.25 μmol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.