Learn More
Thermo Scientific Chemicals May-Gruenwald Stain, pure
Brand: Thermo Scientific Chemicals 194831000
Code : 29
Additional Details : Weight : 0.10000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
May-Gruenwald Stain | |
MFCD00131580 | |
Authentic | |
Pure | |
Brown or Green | |
Fine Crystalline Powder |
524698-43-9 | |
Solubility in water: soluble. | |
in 66:33 IMS:water, 655 to 670nm (blue), 515 to 530nm (red) | |
Glass bottle | |
100 g |
RUO – Research Use Only