Learn More
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
Brand: Thermo Scientific™ K1051
Code : 88
Additional Details : Weight : 3.18000kg
Description
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
Highlights
- Four minute activation time
- High PCR specificity and sensitivity
- Room temperature PCR set up
Specifications
Maxima Hot Start PCR Master Mix (2X) | |
2 x 1.25mL | |
Cloning | |
100 reactions of 50 µl |