Learn More
Thermo Scientific™ Maxima Probe qPCR Master Mixes
Product Code. 11360760 Shop All Thermo Scientific Products
Change view
Click to view available options
:
4 X 12.5mL (for 4000 react. of 25μL)
Product Code.
11360760 4 X 12.5mL (for 4000 react. of 25μL)
1 options
Product Code. 11360760 Supplier Thermo Scientific™ Supplier No. K0263
Optimize qPCR with probe chemistry using standard cycling protocols with these ready-to-use qPCR master mixes.
Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.
Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.
Highlights
  • Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity - detects low copy number targets
  • Wide linear range - accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience - ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY
Specifications
No. of Reactions 4,000 Reactions
Volume 4 X 12.5mL