missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Maxima H Minus Reverse Transcriptase Promotion
Shop All Thermo Scientific Products
Click to view available options
Quantity:
10,000 U
2000 U
Unit Size:
10000 units
2000 units
Description
- Thermostable - 90% active after incubation at 50°C for 60 minutes in a reaction mixture.
- Active up to 65°C
- High yields of full-length cDNA up to 20 kb
- High sensitivity - reproducible cDNA synthesis from a wide range of starting total RNA amounts (1 pg to 5 μg)
- Efficient - complete cDNA synthesis in 15 to 30 minutes
- Increased resistance to common reaction inhibitors
- Incorporates modified nucleotides
Specifications
Specifications
| Concentration | 5X |
| Composition | 250 mM Tris-HCl buffer (pH 8.3 at 25°C), 375 mM KCl, 15 mM MgCl2, 50 mM DTT |
| Content And Storage | -20°C |
| Quantity | 2000 U |
| For Use With (Application) | cDNA Synthesis, RT-PCR, Gene Expression |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction