missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maxi Potassium channel alpha Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Maxi Potassium channel alpha Polyclonal specifically detects Maxi Potassium channel alpha in Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Maxi Potassium channel alpha |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 850-950 of human Maxi Potassium channel alpha (NP_001258447.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?