missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maxi Potassium channel alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | Maxi Potassium channel alpha |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18409451
|
Novus Biologicals
NBP2-33726-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18126244
|
Novus Biologicals
NBP2-33726 |
0.1 mL |
£488.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Maxi Potassium channel alpha Polyclonal specifically detects Maxi Potassium channel alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Maxi Potassium channel alpha | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q12791 | |
| 3778 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: INLCDMCVILSANQNNIDDTSLQDKECILASLNIKSMQFDDSIGVLQANSQGFTPPGMDRSSPDNSPVHGMLRQPSITTGVNIPIITELVNDTNVQFLDQDDDDDPDTELYLTQP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Potassium Channels | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1 | |
| KCNMA1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title