missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas Maxi Cooled Gel Electrophoresis Unit
Click to view available options
Gel Dimensions:
220 x 200 mm
Unit Size:
Each
Description
- Cooled base to cover entire 20 x 20cm gel tray
- Cooled ports to connect the cooled base either to the laboratory water supply or the optional
external chiller - Robust acrylic tank construction 12mm walls, leak proof
- Safety lid with integral power leads
- Shrouded 4mm power output connectors universally compatible with modern-day low to medium voltage power supplies
- Long-life, 99.99% pure platinum electrodes
- Gold-plated terminals with corrosionfree stainless steel washers
- Durable high-impact polystyrene (HIPS) combs
- UV-transparent acrylic gel-casting trays
- Offset asymmetric electrodes
- Buffer recirculation ports
- Colour-coded combs corresponding to 1,1.5 and 2mm thicknesses
Specifications
Specifications
| Type | Gel Electrophoresis Unit |
| Gel Dimensions | 220 x 200 mm |
| Includes | 1 x gel casting tray, 2 x 1.0mm 16 sample combs, power supply connectors and loading strips |
| Model | Maxi cooled |
| Sample Size | 168 |
| Volume (Metric) Buffer | 2200 mL |
| Dimensions (L x W x H) | 47.5 x 27 x 8 cm |
| Length (Metric) Gel | 220 mm |
| Width (Metric) Gel | 220 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
