missing translation for 'onlineSavingsMsg'
Learn More

MAX binding protein Antibody, HRP, Novus Biologicals™

Product Code. 18413871 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413871 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413871 Supplier Novus Biologicals Supplier No. NBP204052H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAX binding protein Polyclonal antibody specifically detects MAX binding protein in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAX binding protein
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias BHLHD3, bHLHd3Max-interacting protein, Class D basic helix-loop-helix protein 3, MAD6, MAX binding protein, MXD6, Myc antagonist MNT, Protein ROX, ROXmax-binding protein MNT
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human MAX Binding Protein using the numbering given in entry NP_064706.1 (GeneID 4335).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4335
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.