missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Max Antibody (103), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90153F
This item is not returnable.
View return policy
Description
Max Monoclonal specifically detects Max in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Max | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 103 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| BHLHD4, bHLHd4MGC11225, bHLHd5, bHLHd6, bHLHd7, bHLHd8, Class D basic helix-loop-helix protein 4, helix-loop-helix zipper protein, MAX protein, MGC10775, MGC18164, MGC34679, MGC36767, MYC associated factor X, Myc-associated factor X, orf1, protein max | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Max (NP_002373; Met1-Ser160). | |
| 0.1 mL | |
| Apoptosis, Phospho Specific | |
| 4149 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction