Learn More
MAVS VISA Mouse anti-Human, Unconjugated, Clone: 1A8E9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66911-1-IG-20UL
 View more versions of this product
Product Code. 16809205
  • £135.00 / 20µL
Description
The MAVS; VISA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MAVS VISA
Monoclonal
900 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence
1A8E9
Unconjugated
20 μL
57506
IgG1