missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAVS Antibody (58N3E1), Novus Biologicals™
Mouse Monoclonal Antibody
£158.00 - £360.00
Specifications
| Antigen | MAVS |
|---|---|
| Clone | 58N3E1 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.1-2 ug/ml |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18117791
|
Novus Biologicals
NBP2-22378SS |
0.025 mg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18147161
|
Novus Biologicals
NBP2-22378 |
0.1 mg |
£360.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Western Blot.Specifications
| MAVS | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Immune System Diseases, Immunology, Protein Kinase | |
| AAH44952.1 | |
| 57506 | |
| Within the range of amino acids 93-298 of human MAVS (isoform 1) were used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 58N3E1 | |
| Western Blot 0.1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051 | |
| MAVS | |
| IgG2b κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title