missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAVS Antibody (58N3B6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80842
This item is not returnable.
View return policy
Description
MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MAVS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 2-5 μg/mL, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin : 3-5 μg/mL | |
| CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 58N3B6 | |
| Unconjugated | |
| PBS | |
| MAVS | |
| Amino acids 93-276 of human MAVS (isoform 1) were used as the immunogen for this antibody. | |
| 0.1 mg | |
| Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase | |
| 57506 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction