missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mavoglurant, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Mavoglurant (AFQ056) is a potent, selective, non-competitive and orally active mGluR5 antagonist, with an IC50 of 30 nM. Mavoglurant shows a >300 fold selectivity for the mGluR5 over all targets (238) tested. Mavoglurant can be used for the research of Fragile X syndrome (FXS), and L-dopa induced dyskinesias in Parkinson's disease.
Specifications
Specifications
| Chemical Name or Material | Mavoglurant |
| CAS | 543906-09-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H23NO3 |
| Quantity | 5 mg |
| Synonym | AFQ056 |
| Solubility Information | DMSO : 120 mg/mL (382.91 mM; Need ultrasonic) |
| SMILES | CC1=CC(C#C[C@]2(O)CCC[C@]3([H])[C@@]2([H])CCN3C(OC)=O)=CC=C1 |
| Molecular Weight (g/mol) | 313.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction