missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mavacamten, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Description
Mavacamten (MYK461) is an orally active modulator of cardiac myosin, with IC50s of 490, 711 nM for bovine cardiac and human cardiac, respectively.
Specifications
Specifications
| Chemical Name or Material | Mavacamten |
| Physical Form | Powder |
| CAS | 1642288-47-8 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H19N3O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | MYK461 SAR439152 |
| Solubility Information | DMSO : 83.33 mg/mL (304.87 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction