Learn More
Invitrogen™ Mature Macrophage Marker Monoclonal Antibody (eBio25F9 (25F9)), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 50-0115-42
Description
Description: The monoclonal antibody 25F9 recognizes a protein on mature macrophages both on the cell surface and in intracellular vesicular structures. Expression is thought to correlate with endocytic macrophages and is useful in many disease models. Expression is not found on immature macrophages or monocytes or any other hematopoietic cell. Applications Reported: This eBio25F9 (25F9) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio25F9 (25F9) antibody has been pre-titrated and tested by flow cytometric analysis of cultured human macrosphages from monocytes. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mature Macrophage Marker Monoclonal antibody specifically detects Mature Macrophage Marker in Human samples. It is validated for Flow Cytometry.
Specifications
| Mature Macrophage Marker | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| Responsibly packaged |
| Flow Cytometry | |
| eBio25F9 (25F9) | |
| eFluor 660 | |
| ED2-like antigen; Macrophages; Mature Macrophages | |
| Affinity chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.