missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ Pipette Tips In Ecotips Refill Packaging System

Product Code. 10591135
Click to view available options
Description:
ECOTIPS
TallTip ECOTIPS
Volume (Metric):
250 μL
1250 μL
Unit Size:
Pack of 960
This item is not returnable. View return policy

Product Code. 10591135

Brand: Thermo Scientific™ 8046

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Process samples quickly with fast rack reloading in an environmentally friendly, space-saving design with this unique tip refill system.

Process samples quickly with fast rack reloading in an environmentally friendly, space-saving design with the Thermo Scientific™ Matrix™ Pipette Tips In Ecotips Refill Packaging System, which reduces plastic waste by eliminating the need for a new tip box for each rack of tips. This system is 100% recyclable and each component is marked with appropriate recycling symbols. Trays of tips are nested in easily accessible packaging that takes up very little space and eliminates the need to dispose of a tip box each time a rack is emptied.

Features:

  • Compact alternative to standard racked tips
  • Quick, efficient refill; reloading empty tip boxes is quick and easy
  • Made from 100% virgin polypropylene; insert trays and boxes come from our own recycled materials
  • Saves bench, shelf and closet space; refills fit easily in a drawer, on a shelf or on the bench.
  • Allows for quick and efficient transfer of tips from packaging to empty tip box
  • Finger tabs on tip trays for easy removal of empty inserts from each tip box
  • Eliminates the need for mechanical towers, clumsy stacks of tips or other transfer mechanisms
TRUSTED_SUSTAINABILITY

Specifications

Description ECOTIPS
Volume (Metric) 1250 μL
Filtered Non-filtered
Sterility Non-sterile
Format Refill
For Use With (Equipment) Thermo Scientific Matrix
Product Line Ecotips, Matrix
Material Virgin Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.