missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MATRIX Orbital Delta Plus

Product Code. 16218681 Shop All IKA Products
Change view
Click to view available options
Orbit:
3 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
16218681 3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16218681 Supplier IKA™ Supplier No. 0030000731
  • ONLINE EXCLUSIVE PRICE
    £2545.00
    £2498.00 / Pack of 1
    Save £47.00
    1% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The all-rounder MATRIX Orbital Delta plus mixes, heats and cools reliably with variable inserts. Whether blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays - reaction vials and plates with even the smallest volumes are reliably mixed and tempered in all laboratory applications.

  • Large and clear display with a convenient menu structure allows for an intuitive operation
  • Firm quick-release fastener for quick and easy changing of the inserts/attachments as well as an automatic attachment detection
  • Quick and effective mixing of liquids with up to 3,000 rpm and 3 mm shaker diameter
  • Reduced waiting time through fast heating and cooling rates of up to 6°C/min (heating) or 2.5°C/min at set-point temperature above the ambient temperature (cooling, 0.5°C/min at set-point temperature between ambient temperature and 15°C below ambient temperature)
  • Two different speed modes for sample-specific temperature control
  • Automatic performance of mixing, heating and cooling tasks through programmable ramps
  • Wide range of inserts for reaction vessels and plates
  • Intelligent speed adaption to insert, attachment and filling volume
  • Aluminum die cast housing ensures secure positioning, excellent stability and everyday robustness
  • No cross-contamination and an optimal mixing result.
TRUSTED_SUSTAINABILITY

Specifications

Orbit 3 mm
Load Bearing Capacity (Metric) 0.3 kg
Speed Control Accuracy 10 rpm Steps
Speed Range 0 to 3000 rpm
Temperature Range (Metric) 1°C to 100°C
Plug Type EU Plug
Temperature Accuracy 0.5°C
Temperature Resolution 1°C
Temperature Uniformity 0.5°C
Frequency 50/60 Hz
Material Aluminum die-cast housing
Dimensions (L x W x H) 351 x 216 x 127 mm
Weight (Metric) 9.5 kg
Display TFT
For Use With (Equipment) 0030001253 MATRIX Cover, 0030001244 MATRIX ThermoCover, 0030000853 MATRIX PCR 96 Attachment, 0030000857 MATRIX PCR 384 Attachment, 0030000840 MATRIX 0.5 mL Attachment, 0030000829 MATRIX 1.5 mL Attachment, 0030000834 MATRIX 2.0 mL Attachment, 0030000859 MATRIX 5 mL Attachment, 0030000844 MATRIX 12 mL Attachment, 0030000821 MATRIX 15 mL Attachment, 0030000863 MATRIX 50 mL Attachment, 0030000848 MATRIX Cryo Attachment
Humidity Range 80%
Includes Power cable; USB cable
Platform Style Exchangable attachments
Timer Range 0 to 6000 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.