missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MATRIX Orbital Delta F0.5

Product Code. 16228681 Shop All IKA Products
Change view
Click to view available options
Orbit:
3 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
16228681 3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16228681 Supplier IKA™ Supplier No. 0030000747
  • ONLINE EXCLUSIVE PRICE
    £2460.00
    £2413.00 / Pack of 1
    Save £47.00
    1% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

IKA MATRIX Orbital Delta F0.5 mixes and heats with a fixed insert for 24 x 0.5 ml tubes. Whether its blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays – samples with even the smallest volumes mix reliably and completely in all laboratory applications.

  • Large and clear display with a convenient menu structure allows for intuitive operation
  • Speeds of up to 2,000 rpm and shaker diameter of 3 mm ensure the liquid mixes quickly and effectively
  • Two different speed modes for gentle heating of the samples
  • Programmable ramps allow for automatic execution of mixing and heating tasks
  • Smart speed adjustment according to insert, attachment and filling volume
  • Aluminum die cast housing ensures secure positioning, excellent stability and everyday robustness.
  • No cross-contamination and an optimal mixing result
TRUSTED_SUSTAINABILITY

Specifications

Orbit 3 mm
Load Bearing Capacity (Metric) 0.3 kg
Speed Control Accuracy 11 rpm Steps
Speed Range 0 to 2000 rpm
Temperature Range (Metric) Ambient +4° to 100°C
Holds 24 x 38 mL Tubes
Tube Capacity 0.5 mL
Plug Type EU Plug
Temperature Accuracy 0.5°C
Temperature Resolution 1°C
Temperature Uniformity 0.5 (at 20°C to 45°C); 1.5°C
Frequency 50/60 Hz
Material Aluminum die-cast housing
Dimensions (L x W x H) 351 x 216 x 149 mm
Weight (Metric) 9.2 kg
Display TFT
Humidity Range 80%
Includes Power cable; USB cable
No. of Positions 24
Platform Style Fixed attachment
Timer Range 0 to 6000 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.