missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matriptase/ST14 Antibody (Oxford Bio patent anti-Matriptase) [Janelia Fluor™ 525], Novus Biologicals™
Description
Matriptase/ST14 Monoclonal antibody specifically detects Matriptase/ST14 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | Matriptase/ST14 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | Oxford Bio patent anti-Matriptase |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein |
| Host Species | Human |
| Immunogen | Matriptase |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?