missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Matriptase/ST14 Antibody (Oxford Bio patent anti-Matriptase) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28073-1mg
This item is not returnable.
View return policy
Description
Matriptase/ST14 Monoclonal antibody specifically detects Matriptase/ST14 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Matriptase/ST14 | |
| Monoclonal | |
| Unconjugated | |
| Q9Y5Y6 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Oxford Bio patent anti-Matriptase | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein | |
| Matriptase | |
| 1 mg | |
| Cancer | |
| 6768 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction