missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matriptase/ST14 Antibody (416802) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3946JF635
This item is not returnable.
View return policy
Description
Matriptase/ST14 Monoclonal antibody specifically detects Matriptase/ST14 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Matriptase/ST14 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein | |
| E. coli-derived recombinant human Matriptase/ST14 Catalytic Domain, Val615-Val855, Accession # Q9Y5Y6 | |
| 0.1 mL | |
| Cancer | |
| 6768 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 416802 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction