missing translation for 'onlineSavingsMsg'
Learn More

Matriptase/ST14 Antibody (2F4), Novus Biologicals™

Product Code. 18368009 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368009 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368009 Supplier Novus Biologicals Supplier No. H00006768M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Matriptase/ST14 Monoclonal antibody specifically detects Matriptase/ST14 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Matriptase/ST14
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2F4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_068813
Gene Alias EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein
Host Species Mouse
Immunogen ST14 (NP_068813, 298 a.a. ~ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PPSYNLTFHSSQNVLLITLITNTERRHPGFEATFFQLPRMSSCGGRLRKAQGTFNSPYYPGHYPPNIDCTWNIEVPNNQHVKVRFKFFYLLEPGVPAGTCPKD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6768
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.