missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matrilin-1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76325CL1
This item is not returnable.
View return policy
Description
Matrilin-1 Polyclonal antibody specifically detects Matrilin-1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Matrilin-1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 4146 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| cartilage matrix protein, matrilin 1, cartilage matrix protein | |
| Antibody was raised against a 12 amino acid synthetic peptide from near the amino terminus of human MATN1. The immunogen is located within amino acids 120 - 170 of MATN1. Amino Acid Squence: EGGRSRSPDISK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction