missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Matrix-3D Plate, 96-well

Product Code. 17279160
Change view
Click to view available options
Color:
Black, Clear
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17279160 Black, Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17279160 Supplier Corning™ Supplier No. 356259

Please to purchase this item. Need a web account? Register with us today!

Corning™ Matrigel™ Matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. It has a high-throughput format that can be used for drug discovery with 3D cell cultures such as organoids, spheroids or tumoroids.

  • A high-throughput pre-coated format that contains Matrigel™ and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids
  • Extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors
  • Each lot is verified for intraplate consistency
  • Tested and found negative for the presence of bacteria and fungi
  • Easy and convenient as Matrigel™ is pre-dispensed into each well
  • Formats provide intra-plate consistency
  • Allows for sandwich/overlay and embedded 3D application workflows
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Breakapart No
Lid With Lid
Content And Storage Store at -20°C
Autoclavable Not Autoclavable
Color Black, Clear
Disposable Single-use
Material Polystyrene
No. of Wells 96
Surface Treatment Matrigel Matrix
Well Shape Flat Bottom (F-well)
Dnase Free No
For Use With (Application) 3D cell culture
Includes Guidelines for Use
Regulatory Status RUO
Rimmed No
RNase Free No
Skirt Color Black with clear bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.