missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Matrix-3D Plate, 384-well
GREENER_CHOICE

Product Code. 17260572
Click to view available options
Color:
Black
White, Clear
No. per Pack:
1
5
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 17260572

Brand: Corning™ 356256

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Corning™ Matrigel™ Matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. It has a high-throughput format that can be used for drug discovery with 3D cell cultures such as organoids, spheroids or tumoroids.

  • A high-throughput pre-coated format that contains Matrigel™ and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids
  • Extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors
  • Each lot is verified for intraplate consistency
  • Tested and found negative for the presence of bacteria and fungi
  • Easy and convenient as Matrigel™ is pre-dispensed into each well
  • Formats provide intra-plate consistency
  • Allows for sandwich/overlay and embedded 3D application workflows
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Breakapart No
Lid With Lid
Content And Storage Store at -20°C
Autoclavable Not Autoclavable
Color Black
Disposable Single-use
Material Polystyrene
No. of Wells 384
Surface Treatment Matrigel Matrix
Well Shape Flat Bottom (F-well)
Dnase Free No
For Use With (Application) 3D cell culture
High-throughput Compatibility Yes
Includes Guidelines for Use
No. per Case 1
No. per Pack 5
Regulatory Status RUO
Rimmed No
RNase Free No
Skirt Color Black with clear bottom
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.