missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Basement Membrane Matrix, LDEV-Free

Product Code. 15575729
Change view
Click to view available options
Quantity:
100 mL (10 mL x 10)
25 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15575729 25 mL Each
15525739 100 mL (10 mL x 10) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15575729 Supplier Corning™ Supplier No. 356232
Quantity Price
1 £1188.96 / Pack of 1
3 £1115.24 / Pack of 1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. Corning™ Matrigel™ Basement Membrane Matrix, LDEV-Free is tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Basement membrane matrix
Without Additives No LDEV
For Use With (Application) Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice
Quantity 25 mL
Source Engelbreth-Holm-Swarm(EHS) Mouse Tumor
Content And Storage Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.